accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH2OOCH3CH3H3C

CAS RN 585-07-9

IUPAC Name: tert-butyl 2-methylprop-2-enoate
Synonyms: T-butyl methacrylate
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: SJMYWORNLPSJQO-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OC(C)(C)C
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results