Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich tert-Butyl methacrylate

Catalog No. 463353250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-335-3250ML 250 mL
46-335-3100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 46-335-3250ML Supplier Sigma Aldrich Supplier No. 463353250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

tert-Butyl methacrylate (tert-BMA) can be used in the formation of homo and block copolymers by atom transfer radical polymerization (ATRP) for potential usage in coatings, biomaterials and flocculants.

Specifications

CAS 585-07-9
Density 0.875 g/mL (at 25°C)
Boiling Point 132°C
Molecular Formula C8H14O2
Refractive Index n20/D 1.415 (literature)
Linear Formula H2C=C(CH3)CO2C(CH3)3
Quantity 250 mL
MDL Number MFCD00048245
Synonym 1,1-Dimethylethyl methacrylate; tert -Butyl alpha-methacrylate; tert -Butyl methacrylate; Acryester TB
Molecular Weight (g/mol) 142.2
Percent Purity 98%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.