CAS RN 585-99-9
IUPAC Name:
6-({[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)oxane-2,3,4,5-tetrol
Synonyms:
polydextrose
Molecular Weight (g/mol):
342.3
Molecular Formula:
C12H22O11
InChi Key:
DLRVVLDZNNYCBX-UHFFFAOYNA-N
SMILES:
OCC1OC(OCC2OC(O)C(O)C(O)C2O)C(O)C(O)C1O
1
–
2
of
2
results
1
alpha-D-(+)-Melibiose, Thermo Scientific Chemicals
CAS: 585-99-9 Molecular Formula: C12H22O11 MDL Number: MFCD00198188 Synonym: 6-O-alpha-D-Galactopyranosyl-D-glucose
2
α-D-(+)-Melibiose Monohydrate, MP Biomedicals™
CAS: 585-99-9 Molecular Formula: C12H22O11·H2O Synonym: 6-O-α-D-galactopyranosyl-D-glucose,α-D-melibiose hydrate