Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-D-(+)-Melibiose, Thermo Scientific Chemicals

Catalog No. AAJ6043906
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6043906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6043906 Supplier Thermo Scientific Chemicals Supplier No. J6043906
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 585-99-9 | Molecular Formula: C12H22O11

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 585-99-9
Molecular Formula C12H22O11
MDL Number MFCD00198188
Synonym 6-O-alpha-D-Galactopyranosyl-D-glucose

Specifications

Color White
Quantity 5 g
Merck Index 14,5820
Solubility Information Soluble in water at 100mg/ml
Formula Weight 342.3
Physical Form Crystalline Powder
Chemical Name or Material alpha-D-(+)-Melibiose
EINECSNumber 209-568-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.