accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNOOHSOOHOSOOHONaNa(E/Z)

CAS RN 5850-63-5

IUPAC Name: 5-hydroxy-3-[2-(naphthalen-1-yl)hydrazin-1-ylidene]-4-oxo-3,4-dihydronaphthalene-2,7-disulfonic acid disodium
Synonyms: 5-hydroxy-3-[2-(naphthalen-1-yl)hydrazin-1-ylidene]-4-oxonaphthalene-2,7-disulfonic acid disodium
Molecular Weight (g/mol): 520.44
Molecular Formula: C20H14N2Na2O8S2
InChi Key: HNMCZESXXQNJMC-UHFFFAOYSA-N
SMILES: [Na].[Na].OC1=C2C(=O)C(=NNC3=CC=CC4=CC=CC=C34)C(=CC2=CC(=C1)S(O)(=O)=O)S(O)(=O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results