Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pontacyl Violet 6R, TCI America™
SDP

Catalog No. P05931G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P05931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P05931G Supplier TCI America Supplier No. P05931G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5850-63-5
Molecular Formula C20H14N2Na2O8S2
Molecular Weight (g/mol) 520.438
MDL Number MFCD00059522
InChI Key HNMCZESXXQNJMC-LSHDGIFOSA-N
PubChem CID 131881523
IUPAC Name (3E)-5-hydroxy-3-(naphthalen-1-ylhydrazinylidene)-4-oxonaphthalene-2,7-disulfonic acid;sodium
SMILES C1=CC=C2C(=C1)C=CC=C2NN=C3C(=CC4=CC(=CC(=C4C3=O)O)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]

Specifications

Color Black-Purple
Quantity 1 g
Formula Weight 518.42
Physical Form Crystalline Powder
Chemical Name or Material Pontacyl Violet 6R
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.