accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOBrBr

CAS RN 58539-11-0

IUPAC Name: ethyl 3-bromo-2-(bromomethyl)propanoate
Synonyms: ethyl 3-bromo-2-(bromomethyl)propanoate
Molecular Weight (g/mol): 273.95
Molecular Formula: C6H10Br2O2
InChi Key: GKSCTYSHDIGNGC-UHFFFAOYSA-N
SMILES: CCOC(=O)C(CBr)CBr
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results