Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-Bromo-2-(bromomethyl)propionate 97.0+%, TCI America™
SDP

Catalog No. E07915G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07915G 5 g
E079125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07915G Supplier TCI America Supplier No. E07915G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 58539-11-0
Molecular Formula C6H10Br2O2
Molecular Weight (g/mol) 273.95
MDL Number MFCD00034154
InChI Key GKSCTYSHDIGNGC-UHFFFAOYSA-N
Synonym 3-Bromo-2-(bromomethyl)propionic Acid Ethyl Ester
PubChem CID 310617
IUPAC Name ethyl 3-bromo-2-(bromomethyl)propanoate
SMILES CCOC(=O)C(CBr)CBr

Specifications

Boiling Point 86°C
Quantity 5 g
Formula Weight 273.95
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Bromo-2-(bromomethyl)propionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.