accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5858-18-4

IUPAC Name: 2,5-dichlorobenzene-1-thiol
Synonyms: 2,5-dichlorobenzenethiol
Molecular Weight (g/mol): 179.06
Molecular Formula: C6H4Cl2S
InChi Key: QIULLHZMZMGGFH-UHFFFAOYSA-N
SMILES: SC1=CC(Cl)=CC=C1Cl
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)