Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichlorobenzenethiol 97.0+%, TCI America™
SDP

Catalog No. D11455G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D11455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D11455G Supplier TCI America Supplier No. D11455G

Chemical Identifiers

CAS 5858-18-4
Molecular Formula C6H4Cl2S
Molecular Weight (g/mol) 179.058
MDL Number MFCD00004831
InChI Key QIULLHZMZMGGFH-UHFFFAOYSA-N
Synonym 2,5-dichlorothiophenol, 2,5-dichlorobenzene-1-thiol, benzenethiol, 2,5-dichloro, 2,5-dichloro thiophenol, thiophenol, 2,5-dichloro, 2,5-dichlorophenylmercaptan, pubchem6801, thiophenol,5-dichloro, 2,5-dichlorobenzenthiol, 2,5-dichloro-thiophenol
PubChem CID 79965
IUPAC Name 2,5-dichlorobenzenethiol
SMILES C1=CC(=C(C=C1Cl)S)Cl

Specifications

Color White-Yellow
Boiling Point 116°C
Quantity 5 g
Formula Weight 179.06
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,5-Dichlorobenzenethiol
EINECSNumber (3)-2898
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.