accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOO(E)

CAS RN 58598-42-8

IUPAC Name: 3-[(10E)-docos-10-en-1-yl]oxolane-2,5-dione
Synonyms: 3-[(10E)-docos-10-en-1-yl]oxolane-2,5-dione
Molecular Weight (g/mol): 406.65
Molecular Formula: C26H46O3
InChi Key: PYGZRGQVVVNPHG-OUKQBFOZNA-N
SMILES: CCCCCCCCCCC\C=C\CCCCCCCCCC1CC(=O)OC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results