accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 586-35-6

IUPAC Name: 2-bromobenzene-1,4-dicarboxylic acid
Synonyms: 2-bromobenzene-1,4-dicarboxylic acid
Molecular Weight (g/mol): 245.03
Molecular Formula: C8H5BrO4
InChi Key: QPBGNSFASPVGTP-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C(O)=O)C(Br)=C1
Molecular Weight (g/mol) 
  • (7)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (4)
  • (4)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (6)
  • (7)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (4)
  • (4)
  • (2)
  • (2)