Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromoterephthalic Acid 99%, Thermo Scientific™

Catalog No. AC151730250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC151730250 25 g
AC151730050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC151730250 Supplier Thermo Scientific Chemicals Supplier No. 151730250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 586-35-6
Molecular Formula C8H5BrO4
Molecular Weight (g/mol) 245.028
InChI Key QPBGNSFASPVGTP-UHFFFAOYSA-N
Synonym bromoterephthalic acid, 2-bromobenzene-1,4-dicarboxylic acid, 2-bromoterephthalicacid, 2-bromo-terephthalic acid, 2-bromo-1,4-benzenedicarboxylic acid, 1,4-benzenedicarboxylic acid, 2-bromo, labotest-bb lt00454996, pubchem12798, 2-bromo terephthalic acid, acmc-1aoy9
PubChem CID 68513
IUPAC Name 2-bromoterephthalic acid
SMILES C1=CC(=C(C=C1C(=O)O)Br)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Linear Formula BrC6H3(CO2H)2
Quantity 25 g
Beilstein 09, 848
Formula Weight 245.03
Percent Purity ≥98.5%
Physical Form Powder
Chemical Name or Material 2-Bromoterephthalic acid, 99%
Show More Show Less
EINECSNumber 209-572-8
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.