accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOO

CAS RN 58605-10-0

IUPAC Name: methyl 3,5-bis(benzyloxy)benzoate
Synonyms: methyl 3,5-bis(benzyloxy)benzoate
Molecular Weight (g/mol): 348.4
Molecular Formula: C22H20O4
InChi Key: GBQCMRLPXFXVIN-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(OCC2=CC=CC=C2)=CC(OCC2=CC=CC=C2)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results