Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3,5-Dibenzyloxybenzoate 98.0+%, TCI America™
SDP

Catalog No. B20925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20925G 5 g
B209225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20925G Supplier TCI America Supplier No. B20925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 58605-10-0
Molecular Formula C22H20O4
Molecular Weight (g/mol) 348.398
MDL Number MFCD00157205
InChI Key GBQCMRLPXFXVIN-UHFFFAOYSA-N
Synonym 3,5-Bis(benzyloxy)benzoic Acid Methyl Ester, 3,5-Dibenzyloxybenzoic Acid Methyl Ester
PubChem CID 2733652
IUPAC Name methyl 3,5-bis(phenylmethoxy)benzoate
SMILES COC(=O)C1=CC(=CC(=C1)OCC2=CC=CC=C2)OCC3=CC=CC=C3

Specifications

Melting Point 71°C
Color White-Yellow
Quantity 5 g
Formula Weight 348.40
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3,5-Dibenzyloxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.