accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5867-45-8

IUPAC Name: N-(pyridin-3-yl)acetamide
Synonyms: 3-acetaminopyridine
Molecular Weight (g/mol): 136.15
Molecular Formula: C7H8N2O
InChi Key: JVYIBLHBCPSTKF-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CN=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results