Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Acetamidopyridine 98.0+%, TCI America™
SDP

Catalog No. A19125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A19125G Supplier TCI America Supplier No. A19125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5867-45-8
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.154
InChI Key JVYIBLHBCPSTKF-UHFFFAOYSA-N
PubChem CID 79978
IUPAC Name N-pyridin-3-ylacetamide
SMILES CC(=O)NC1=CN=CC=C1

Specifications

Melting Point 136°C
Color White
Boiling Point 327°C
Quantity 5 g
Formula Weight 136.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Acetamidopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.