accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOONOONOONN

CAS RN 5868-05-3

IUPAC Name: 3-(pyridine-3-carbonyloxy)-2,2-bis[(pyridine-3-carbonyloxy)methyl]propyl pyridine-3-carboxylate
Synonyms: bufor niceritrol perycit pentaerythritoltetranicotinate cardiolipol niceritrolum SK 1 (pharmaceutical) pentaerythritol tetranicotinate tetranicotinate, pentaerythritol neopentanetetrayl ester nicotinic acid
Molecular Weight (g/mol): 556.53
Molecular Formula: C29H24N4O8
InChi Key: KUEUWHJGRZKESU-UHFFFAOYSA-N
SMILES: O=C(OCC(COC(=O)C1=CC=CN=C1)(COC(=O)C1=CC=CN=C1)COC(=O)C1=CC=CN=C1)C1=CC=CN=C1
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results