Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niceritrol 98.0+%, TCI America™
SDP

Catalog No. N097425MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N097425MG 25 mg
N0974100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N097425MG Supplier TCI America Supplier No. N097425MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 5868-05-3 | C29H24N4O8 | 556.53 g/mol

Chemical Identifiers

CAS 5868-05-3
Molecular Formula C29H24N4O8
Molecular Weight (g/mol) 556.53
MDL Number MFCD00215982
InChI Key KUEUWHJGRZKESU-UHFFFAOYSA-N
Synonym Pentaerythritol Tetranicotinate
PubChem CID 4476
IUPAC Name [3-(pyridine-3-carbonyloxy)-2,2-bis(pyridine-3-carbonyloxymethyl)propyl] pyridine-3-carboxylate
SMILES C1=CC(=CN=C1)C(=O)OCC(COC(=O)C2=CN=CC=C2)(COC(=O)C3=CN=CC=C3)COC(=O)C4=CN=CC=C4

Specifications

Melting Point 165°C
Color White-Yellow
Quantity 25 mg
Percent Purity ≥98.0% (HPLC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Niceritrol
RTECSNumber QT1980000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.