accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2N

CAS RN 58728-64-6

IUPAC Name: 4-aminonaphthalene-1-carbonitrile
Synonyms: 4-aminonaphthalene-1-carbonitrile
Molecular Weight (g/mol): 168.2
Molecular Formula: C11H8N2
InChi Key: LUSIZUFVMKYWGX-UHFFFAOYSA-N
SMILES: NC1=C2C=CC=CC2=C(C=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)