accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5873-51-8

IUPAC Name: 3-[1-(7-chloro-3-ethyl-4-oxo-3,4-dihydroquinazolin-2-yl)propyl]-1-(3-chlorophenyl)-3-propylurea
Synonyms: 3-[1-(7-chloro-3-ethyl-4-oxoquinazolin-2-yl)propyl]-1-(3-chlorophenyl)-3-propylurea
Molecular Weight (g/mol): 461.39
Molecular Formula: C23H26Cl2N4O2
InChi Key: FYNTWOYQGCUHQF-UHFFFAOYNA-N
SMILES: CCCN(C(CC)C1=NC2=CC(Cl)=CC=C2C(=O)N1CC)C(=O)NC1=CC=CC(Cl)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results