Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-aminophenyl) Sulfide 98.0+%, TCI America™
SDP

Catalog No. B292825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B292825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B292825G Supplier TCI America Supplier No. B292825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5873-51-8
Molecular Formula C23H26Cl2N4O2
Molecular Weight (g/mol) 461.39
MDL Number MFCD03094662
InChI Key FYNTWOYQGCUHQF-UHFFFAOYNA-N
Synonym 2-Aminophenyl Sulfide, 2,2′C-Diaminodiphenyl Sulfide, 2,2′C-Thiodianiline
PubChem CID 612177
IUPAC Name 3-[1-(7-chloro-3-ethyl-4-oxo-3,4-dihydroquinazolin-2-yl)propyl]-1-(3-chlorophenyl)-3-propylurea
SMILES CCCN(C(CC)C1=NC2=CC(Cl)=CC=C2C(=O)N1CC)C(=O)NC1=CC=CC(Cl)=C1

Specifications

Melting Point 87°C
Quantity 25 g
Formula Weight 216.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2-aminophenyl) Sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.