accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 58743-75-2

IUPAC Name: 4'-ethyl-[1,1'-biphenyl]-4-carbonitrile
Synonyms: 4'-ethyl-[1,1'-biphenyl]-4-carbonitrile
Molecular Weight (g/mol): 207.28
Molecular Formula: C15H13N
InChi Key: DLLIPJSMDJCZRF-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C=C1)C1=CC=C(C=C1)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results