Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Cyano-4'-ethylbiphenyl 98.0+%, TCI America™
SDP

Catalog No. C28925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C28925G Supplier TCI America Supplier No. C28925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 58743-75-2
Molecular Formula C15H13N
Molecular Weight (g/mol) 207.276
MDL Number MFCD00799420
InChI Key DLLIPJSMDJCZRF-UHFFFAOYSA-N
Synonym 4-cyano-4'-ethylbiphenyl, 4'-ethyl-1,1'-biphenyl-4-carbonitrile, 4-ethyl-4'-cyanobiphenyl, 4-4-ethylphenyl benzonitrile, 1,1'-biphenyl-4-carbonitrile, 4'-ethyl, 4'-ethylbiphenyl-4-carbonitrile, 4-4-ethylphenyl benzenecarbonitrile, pubchem2521, 4'-ethyl-4-cyanobiphenyl
PubChem CID 93890
IUPAC Name 4-(4-ethylphenyl)benzonitrile
SMILES CCC1=CC=C(C=C1)C2=CC=C(C=C2)C#N

Specifications

Melting Point 77°C
Color White
Boiling Point 205°C
Quantity 5 g
Formula Weight 207.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-4′-ethylbiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.