accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 58757-38-3

IUPAC Name: 6-chloropyridine-3-carbonyl chloride
Synonyms: 6-chloropyridine-3-carbonyl chloride
Molecular Weight (g/mol): 176
Molecular Formula: C6H3Cl2NO
InChi Key: FMEBIWNKYZUWFV-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=C(Cl)N=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results