Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloronicotinoyl Chloride 98.0+%, TCI America™
SDP

Catalog No. C17265G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17265G 5 g
C172625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17265G Supplier TCI America Supplier No. C17265G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 58757-38-3
Molecular Formula C6H3Cl2NO
Molecular Weight (g/mol) 175.996
MDL Number MFCD00051775
InChI Key FMEBIWNKYZUWFV-UHFFFAOYSA-N
Synonym 6-chloronicotinoyl chloride, 6-chloronicotinoylchloride, 2-chloropyridine-5-carbonyl chloride, 3-pyridinecarbonylchloride, 6-chloro, 3-pyridinecarbonyl chloride, 6-chloro, 6-chloro-nicotinoyl chloride, pubchem18665, 6-chloronicotinic chloride, acmc-1ay8e, 6-chloro-nicotinic chloride
PubChem CID 2782165
IUPAC Name 6-chloropyridine-3-carbonyl chloride
SMILES C1=CC(=NC=C1C(=O)Cl)Cl

Specifications

Melting Point 51°C
Boiling Point 85°C
Quantity 5 g
UN Number 3261
Formula Weight 176.00
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloronicotinoyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.