CAS RN 587852-28-6
CAS RN 587852-28-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ PIM1/2 Kinase Inhibitor VI
The PIM1/2 Kinase Inhibitor VI, also referenced under CAS 587852-28-6, controls the biological activity of PIM1/2.