accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 588-23-8

IUPAC Name: N-{2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]phenyl}acetamide
Synonyms: N-{2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]phenyl}acetamide
Molecular Weight (g/mol): 267.33
Molecular Formula: C16H17N3O
InChi Key: GFMVVEHDEIYWTL-UHFFFAOYSA-N
SMILES: CC(=O)NC1=C(C)C=C(C=C1)N=NC1=CC=CC=C1C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results