Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetamido-2',3-dimethylazobenzene 98.0+%, TCI America™
SDP

Catalog No. A007325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A007325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A007325G Supplier TCI America Supplier No. A007325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 588-23-8
Molecular Formula C16H17N3O
Molecular Weight (g/mol) 267.33
MDL Number MFCD00059332
InChI Key GFMVVEHDEIYWTL-UHFFFAOYSA-N
Synonym o-Acetamidoazotoluene, N-Acetyl-o-aminoazotoluene, 4′C-o-Tolylazo-o-acetotoluidine
PubChem CID 562757
IUPAC Name N-{2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]phenyl}acetamide
SMILES CC(=O)NC1=C(C)C=C(C=C1)N=NC1=CC=CC=C1C

Specifications

Melting Point 187°C
Quantity 25 g
Formula Weight 267.33
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetamido-2′,3-dimethylazobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.