accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 588-67-0

IUPAC Name: (butoxymethyl)benzene
Synonyms: benzene, (butoxymethyl)-
Molecular Weight (g/mol): 164.25
Molecular Formula: C11H16O
InChi Key: MAYUYFCAPVDYBQ-UHFFFAOYSA-N
SMILES: CCCCOCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results