Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Butyl Ether 95.0+%, TCI America™
SDP

Catalog No. B02395G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B02395G 5 g
B023925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B02395G Supplier TCI America Supplier No. B02395G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 588-67-0
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.25
MDL Number MFCD00039954
InChI Key MAYUYFCAPVDYBQ-UHFFFAOYSA-N
Synonym benzyl butyl ether, ether, benzyl butyl, butyl benzyl ether, benzyl n-butyl ether, benzene, butoxymethyl, butoxymethyl benzene, n-butyl benzyl ether, unii-8483x8zo8n, ether, benzyl n-butyl, butoxyphenylmethane
PubChem CID 61134
IUPAC Name (butoxymethyl)benzene
SMILES CCCCOCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 220°C
Quantity 5 g
Formula Weight 164.25
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Butyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.