accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OBr

CAS RN 588-96-5

IUPAC Name: 1-bromo-4-ethoxybenzene
Synonyms: benzene, 1-bromo-4-ethoxy-
Molecular Weight (g/mol): 201.06
Molecular Formula: C8H9BrO
InChi Key: WVUYYXUATWMVIT-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(Br)C=C1
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)