Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromophenetole 98.0+%, TCI America™
SDP

Catalog No. B143425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B143425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B143425G Supplier TCI America Supplier No. B143425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 588-96-5
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.063
MDL Number MFCD00000098
InChI Key WVUYYXUATWMVIT-UHFFFAOYSA-N
Synonym 4-bromophenetole, p-bromophenetole, benzene, 1-bromo-4-ethoxy, p-bromoethoxybenzene, p-ethoxybromobenzene, p-ethoxyphenyl bromide, p-bromophenol ethyl ether, 4-bromophenyl ethyl ether, 1-bromo-4-ethoxy-benzene, 4-bromoethoxybenzene
PubChem CID 68523
IUPAC Name 1-bromo-4-ethoxybenzene
SMILES CCOC1=CC=C(C=C1)Br

Specifications

Melting Point 4°C
Color Yellow
Boiling Point 233°C
Quantity 25 g
Formula Weight 201.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromophenetole
EINECSNumber (3)-0845
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.