CAS RN 58846-77-8
CAS RN 58846-77-8
IUPAC Name:
(2R,3R,4S,5S,6R)-2-(decyloxy)-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms:
decyl glucoside
Molecular Weight (g/mol):
320.43
Molecular Formula:
C16H32O6
InChi Key:
JDRSMPFHFNXQRB-IBEHDNSVSA-N
SMILES:
CCCCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O
1
–
1
of
1
results
1
Decyl beta-D-glucopyranoside, Thermo Scientific Chemicals
CAS: 58846-77-8 Molecular Formula: C16H32O6 Molecular Weight (g/mol): 320.426 MDL Number: MFCD00063297 InChI Key: JDRSMPFHFNXQRB-IBEHDNSVSA-N Synonym: decyl beta-d-glucopyranoside,n-decyl-b-d-glucopyranoside,decyl-beta-d-glucopyranoside,n-decyl-beta-d-glucopyranoside,unii-z17h97ea6y,n-decyl glucoside,decyl beta-d-glucoside,triton x 190,28-deacetylsendanin,beta-d-glucoside, decyl PubChem CID: 62142 IUPAC Name: (2R,3R,4S,5S,6R)-2-decoxy-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: CCCCCCCCCCOC1C(C(C(C(O1)CO)O)O)O