accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONH

CAS RN 58869-89-9

IUPAC Name: decahydroquinolin-4-one
Synonyms: octahydro-1H-quinolin-4-one
Molecular Weight (g/mol): 153.23
Molecular Formula: C9H15NO
InChi Key: VZTBFFGJFFYFRT-UHFFFAOYNA-N
SMILES: O=C1CCNC2CCCCC12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results