Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octahydro-4(1H)-quinolinone, 97%, Thermo Scientific™

Catalog No. AAH5018003
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5018003 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5018003 Supplier Thermo Scientific Chemicals Supplier No. H5018003

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Octahydro-4(1H)-quinolinone is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 58869-89-9
Molecular Formula C9H15NO
Quantity 1g
MDL Number MFCD02734410
Synonym octahydro-4 1h-quinolinone, octahydroquinolin-4 1h-one, octahydro-quinolin-4-one, octahydro-1h-quinolin-4-one, decahydro-4-quinolone, decahydroquinolin-4-one, octahydroquinolin-4-one, acmc-20ak0e, octahydroquinoline-4 1h-one, 4 1h-quinolinone, octahydro-, hydrochloride
InChI Key VZTBFFGJFFYFRT-UHFFFAOYSA-N
SMILES C1CCC2C(C1)C(=O)CCN2
IUPAC Name 2,3,4a,5,6,7,8,8a-octahydro-1H-quinolin-4-one
Molecular Weight (g/mol) 153.225
PubChem CID 3143020
Formula Weight 153.23
Percent Purity 97%
Chemical Name or Material Octahydro-4(1H)-quinolinone
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.