accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOHHO

CAS RN 58871-05-9

IUPAC Name: (3,4-dihydroxy-3,4-dihydro-2H-pyran-2-yl)methyl benzoate
Synonyms: (3,4-dihydroxy-3,4-dihydro-2H-pyran-2-yl)methyl benzoate
Molecular Weight (g/mol): 250.25
Molecular Formula: C13H14O5
InChi Key: DIEYDTYJOXFFJG-UHFFFAOYNA-N
SMILES: OC1C=COC(COC(=O)C2=CC=CC=C2)C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results