Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-O-Benzoyl-D-glucal, 97%, Thermo Scientific™

Catalog No. AAH60465MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60465MD 250 mg
AAH6046503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH60465MD Supplier Thermo Scientific Chemicals Supplier No. H60465MD

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58871-05-9
Molecular Formula C13H14O5
Molecular Weight (g/mol) 250.25
MDL Number MFCD01321275
InChI Key DIEYDTYJOXFFJG-UTUOFQBUSA-N
Synonym 6-o-benzoyl-d-glucal, 2r,3s,4r-3,4-dihydroxy-3,4-dihydro-2h-pyran-2-yl methyl benzoate, 6-o-benzoylglucal, d-arabino-hex-1-enitol, 1,5-anhydro-2-deoxy-, 6-benzoate
PubChem CID 14308105
IUPAC Name [(2R,3S,4R)-3,4-dihydroxy-3,4-dihydro-2H-pyran-2-yl]methyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OCC2C(C(C=CO2)O)O

Specifications

Melting Point 106°C to 111°C
Quantity 250 mg
Optical Rotation +44° (c=1.4 in Chloroform)
Formula Weight 250.25
Percent Purity 97%
Chemical Name or Material 6-O-Benzoyl-D-glucal
Recommended Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.