accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 589-37-7

IUPAC Name: pentane-1,3-diamine
Synonyms: pentane-1,3-diamine
Molecular Weight (g/mol): 102.18
Molecular Formula: C5H14N2
InChi Key: WTSXICLFTPPDTL-UHFFFAOYNA-N
SMILES: CCC(N)CCN
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)