Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diaminopentane 96.0+%, TCI America™
SDP

Catalog No. D2252500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2252500ML 500 mL
D225225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2252500ML Supplier TCI America Supplier No. D2252500ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 589-37-7
Molecular Formula C5H14N2
Molecular Weight (g/mol) 102.181
MDL Number MFCD00134550
InChI Key WTSXICLFTPPDTL-UHFFFAOYSA-N
Synonym 1,3-Pentanediamine
PubChem CID 102444
IUPAC Name pentane-1,3-diamine
SMILES CCC(CCN)N

Specifications

Color Yellow
Boiling Point 164°C
Quantity 500 mL
UN Number 2734
Formula Weight 102.18
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Diaminopentane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.