accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONHOH

CAS RN 589-41-3

IUPAC Name: ethyl N-hydroxycarbamate
Synonyms: N-hydroxyurethane
Molecular Weight (g/mol): 105.09
Molecular Formula: C3H7NO3
InChi Key: VGEWEGHHYWGXGG-UHFFFAOYSA-N
SMILES: CCOC(=O)NO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results