accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOCH3CH3

CAS RN 589-59-3

IUPAC Name: 2-methylpropyl 3-methylbutanoate
Synonyms: isobutyl isovalerate
Molecular Weight (g/mol): 158.24
Molecular Formula: C9H18O2
InChi Key: KEBDNKNVCHQIJU-UHFFFAOYSA-N
SMILES: CC(C)COC(=O)CC(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results