accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3CH3(Z)

CAS RN 589-66-2

IUPAC Name: 2-methylpropyl (2Z)-but-2-enoate
Synonyms: 2-methylpropyl (2Z)-but-2-enoate
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: XDOWKOALJBOBBL-PLNGDYQASA-N
SMILES: C\C=C/C(=O)OCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results