Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyl Crotonate 98.0+%, TCI America™
SDP

Catalog No. C129525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C129525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C129525ML Supplier TCI America Supplier No. C129525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 589-66-2
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00036651
InChI Key XDOWKOALJBOBBL-PLNGDYQASA-N
Synonym Crotonic Acid Isobutyl Ester, Isobutyl 2-Butenoate, 2-Butenoic Acid Isobuyl Ester
PubChem CID 6435824
IUPAC Name 2-methylpropyl (2Z)-but-2-enoate
SMILES C\C=C/C(=O)OCC(C)C

Specifications

Color Colorless
Boiling Point 171°C
Quantity 25 mL
UN Number 3272
Formula Weight 142.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Crotonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.