accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NHOS(E)

CAS RN 58905-71-8

IUPAC Name: (E)-N'-hydroxythiophene-3-carboximidamide
Synonyms: (E)-N'-hydroxythiophene-3-carboximidamide
Molecular Weight (g/mol): 142.18
Molecular Formula: C5H6N2OS
InChi Key: KLQUDAATMCQZEF-UHFFFAOYSA-N
SMILES: N\C(=N\O)C1=CSC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results