accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3COOHOONHOO

CAS RN 5891-45-2

IUPAC Name: 4-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoic acid
Synonyms: 4-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoic acid
Molecular Weight (g/mol): 337.37
Molecular Formula: C17H23NO6
InChi Key: VJECGKAFPHEJQS-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)C(CCC(O)=O)NC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)