Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-tert-Butyl N-Carbobenzoxy-L-glutamate 98.0+%, TCI America™
SDP

Catalog No. B49125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49125G 5 g
B49121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49125G Supplier TCI America Supplier No. B49125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5891-45-2
Molecular Formula C17H23NO6
Molecular Weight (g/mol) 337.37
MDL Number MFCD00077016
InChI Key VJECGKAFPHEJQS-UHFFFAOYNA-N
Synonym z-glu-otbu, s-4-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, s-4-benzyloxycarbonylamino-5-tert-butoxy-5-oxopentanoic acid, cbz-glu-otbu, z-glu-o-tbu, pubchem18088, pubchem18089, 1-tert-butyl n-cbz-l-glutamate, l-glutamic acid, n-phenylmethoxy carbonyl-, 1-1,1-dimethylethyl ester, z-l-glutamic acid, a-tert.butyl ester
PubChem CID 6994036
IUPAC Name 4-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)C(CCC(O)=O)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 83°C
Color White
Quantity 5 g
Formula Weight 337.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-tert-Butyl N-Carbobenzoxy-L-glutamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.