accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH(R)NHOO

CAS RN 58917-85-4

IUPAC Name: benzyl N-[(2R)-1-hydroxy-3-phenylpropan-2-yl]carbamate
Synonyms: benzyl N-[(2R)-1-hydroxy-3-phenylpropan-2-yl]carbamate
Molecular Weight (g/mol): 285.34
Molecular Formula: C17H19NO3
InChi Key: WPOFMMJJCPZPAO-MRXNPFEDSA-N
SMILES: OC[C@@H](CC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)