Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-D-phenylalaninol 97.0+%, TCI America™
SDP

Catalog No. C16095G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16095G 5 g
C16091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C16095G Supplier TCI America Supplier No. C16095G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 58917-85-4
Molecular Formula C17H19NO3
Molecular Weight (g/mol) 285.34
MDL Number MFCD00191193
InChI Key WPOFMMJJCPZPAO-MRXNPFEDSA-N
Synonym cbz-d-phenylalaninol, z-d-phenylalaninol, n-carbobenzoxy-d-phenylalaninol, r-+-2-cbz-amino-3-phenyl-1-propanol, r-+-2-benzyloxycarbonylamino-3-phenyl-1-propanol, n-carbobenzyloxy-d-phenylalaninol, r-+-2-carbobenzyloxyamino-3-phenyl-1-propanol, benzyl n-2r-1-hydroxy-3-phenylpropan-2-yl carbamate, r-2-z-amino-3-phenyl-1-propanol, benzyl 2r-1-hydroxy-3-phenylpropan-2-yl carbamate
PubChem CID 736170
IUPAC Name benzyl N-[(2R)-1-hydroxy-3-phenylpropan-2-yl]carbamate
SMILES OC[C@@H](CC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 93°C
Color Yellow
Quantity 5 g
Formula Weight 285.34
Percent Purity ≥97.0% (HPLC)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-D-phenylalaninol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.