accessibility menu, dialog, popup

UserName

Viewing profile as user:
EuEuOOOOOOOOO

CAS RN 5895-48-7

IUPAC Name: dieuropium(3+) tricarbonate
Synonyms: dieuropium(3+) tricarbonate
Molecular Weight (g/mol): 483.95
Molecular Formula: C3Eu2O9
InChi Key: MBFNJTXDYDGXDL-UHFFFAOYSA-H
SMILES: [Eu+3].[Eu+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results